Skip to Content
Merck
All Photos(1)

Key Documents

H6291

Sigma-Aldrich

IL-1β human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Synonym(s):

Interleukin-1β human, Lymphocyte activating factor, IL-1β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Interleukin-1β human, IL-1β, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture, endotoxin tested

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.05 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 18-25 kDa (glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL1B(3553)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Interleukin-1 (IL-1), originally known as Lymphocyte Activating Factor (LAF), activates T cell lymphocytes, which then proliferate and secrete interleukin-2 (IL-2). IL-1 is released primarily from stimulated macrophages and monocytes, but has also been shown to be released from several other types and is thought to play a key role in inflammatory and immune responses. Other synonyms for IL-1 include: endogenous pyrogen (EP), mitogenic protein (MP), Helper Peak-1 (HP-1), T Cell Replacing Factor III (TRF III or TRFM), B Cell Activating Factor (BAF) and B Cell Differentiation Factor (BDF) The two closely related agents Interleukin-1α (IL-1α) and Interleukin-1β (IL-1β) share 62% homology in amino acid sequence and elicit nearly identical biological responses. IL-1α and IL-1β are both approximately 17 kDa with some heterogeneity in the amount of glycosylation.
Interleukin-1 activates T cell lymphocytes, which then proliferate and secrete interleukin-2 (IL-2). IL-1 is released primarily from stimulated macrophages and monocytes, but has also been shown to be released from several other types and is thought to play a key role in inflammatory and immune responses.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine Recombinant Human IL-1β, expressed in human HEK 293 cells, is a glycosylated monomer with a molecular mass of 18-25 kDa. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The activity is determined by the dose-dependent stimulation of the proliferation of mouse D10S cells.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J O R Hernandez et al.
Nature communications, 12(1), 6496-6496 (2021-11-13)
The lack of animal models for some human diseases precludes our understanding of disease mechanisms and our ability to test prospective therapies in vivo. Generation of kidney organoids from Tuberous Sclerosis Complex (TSC) patient-derived-hiPSCs allows us to recapitulate a rare
Charles A Dinarello et al.
Current opinion in endocrinology, diabetes, and obesity, 17(4), 314-321 (2010-07-01)
To understand the role of inflammation as the fundamental cause of type 2 diabetes and specifically to examine the contribution of IL-1beta. Recent studies from animals, in-vitro cultures and clinical trials provide evidence that support a causative role for IL-1beta
Daniela Guidone et al.
JCI insight (2022-10-12)
The fluid covering the surface of airway epithelia represents a first barrier against pathogens. The chemical and physical properties of the airway surface fluid are controlled by the activity of ion channels and transporters. In cystic fibrosis (CF), loss of

Articles

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service