Skip to Content
Merck
All Photos(1)

Documents

H6024

Sigma-Aldrich

Hoechst Stain solution

Synonym(s):

Bisbenzimide H 33258

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

form

solution

application(s)

histology

storage temp.

2-8°C

SMILES string

O.Cl.Cl.Cl.CN1CCN(CC1)c2ccc3NC(=NCc3c2)c4ccc5NC(=NCc5c4)c6ccc(O)cc6

InChI

1S/C27H28N6O.3ClH.H2O/c1-32-10-12-33(13-11-32)22-5-9-25-21(15-22)17-29-27(31-25)19-4-8-24-20(14-19)16-28-26(30-24)18-2-6-23(34)7-3-18;;;;/h2-9,14-15,34H,10-13,16-17H2,1H3,(H,28,30)(H,29,31);3*1H;1H2

InChI key

OWRSPPSBNWJJAR-UHFFFAOYSA-N

General description

Hoechst stains are part of a family of blue fluorescent dyes commonly used to stain DNA. It gets incorporated biosynthetically into interphase nuclei in fixed cytologic preparations and in unfixed cultured cells. It can also be used to detect biosynthetic incorporation of 5-bromodeoxyuridine into the DNA of living cells. Hoechst stains property may allow fluorescence-activated cell and chromosome sorting according to the extent of DNA synthesis which may provide a bridge between biochemical and cytologic analyses of processes related to DNA replication.

Application

A fluorescent DNA stain qualified for use in Mycoplama staining.

Components

Contains 0.5μg/ml Hoechst bisbenzimide 33258 fluorochrome stain and thimerosal.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Latt et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 23(7), 493-505 (1975-07-01)
A number of applications of the detection of deoxyribonucleic acid synthesis by fluorescence microscopy are illustrated. These include (a) the analysis of sister chromatid exchanges and sister chromatid segregation at mitosis, (b) the location of chromosome regions containing deoxyribonucleic acid
Nicola Cirillo et al.
British journal of cancer, 117(7), 984-993 (2017-08-11)
Recent studies have shown that production of cortisol not only takes place in several non-adrenal peripheral tissues such as epithelial cells but, also, the local inter-conversion between cortisone and cortisol is regulated by the 11β-hydroxysteroid dehydrogenases (11β-HSDs). However, little is
Georg Tascher et al.
Cells, 8(2) (2019-02-24)
Of the hepatic cell lines developed for in vitro studies of hepatic functions as alternatives to primary human hepatocytes, many have lost major liver-like functions, but not HepaRG cells. The increasing use of the latter worldwide raises the need for
Marisa Müller et al.
PLoS biology, 9(4), e1000611-e1000611 (2011-04-29)
In eukaryotes, hundreds of mRNAs are localized by specialized transport complexes. For localization, transcripts are recognized by RNA-binding proteins and incorporated into motor-containing messenger ribonucleoprotein particles (mRNPs). To date, the molecular assembly of such mRNPs is not well understood and
Yifat Ofir-Birin et al.
Frontiers in immunology, 9, 1011-1011 (2018-06-09)
Extracellular vesicles are essential for long distance cell-cell communication. They function as carriers of different compounds, including proteins, lipids and nucleic acids. Pathogens, like malaria parasites (Plasmodium falciparum, Pf), excel in employing vesicle release to mediate cell communication in diverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service