Skip to Content
Merck
All Photos(1)

Key Documents

H5263

Sigma-Aldrich

Heparin-Acrylic beads

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
13111000

Pricing and availability is not currently available.

biological source

heparin from Porcine intestinal mucosa

form

saline suspension

extent of labeling

0.5-1.5 mg per mL (activity approx. 170 USP units per mg)

matrix

acrylic beads

matrix activation

oxirane

matrix spacer

5 atoms

storage temp.

2-8°C

Physical form

Suspension in 0.15 M NaCl containing 0.02% thimerosal

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quliang Gu et al.
Experimental cell research, 333(2), 261-272 (2015-04-01)
Glipizide, a second-generation sulfonylurea, has been widely used for the treatment of type 2 diabetes. However, it is controversial whether or not glipizide would affect angiogenesis or vasculogenesis. In the present study, we used early chick embryo model to investigate
Muhammad Abu-Elmagd et al.
BMC medical genomics, 8 Suppl 1, S8-S8 (2015-03-19)
Negative regulators of signal transduction cascades play critical roles in controlling different aspects of normal embryonic development. Sprouty2 (Spry2) negatively regulates receptor tyrosine kinases (RTK) and FGF signalling and is important in differentiation, cell migration and proliferation. In vertebrate embryos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service