H4917
Cytochrome P450 3A5 human
recombinant, expressed in E. coli
Synonym(s):
Cytochrome P450, purified, soluble, CYP3A5
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
description
purified, soluble
form
liquid
specific activity
12-100 units/mg protein
mol wt
56998.2 g/mol
concentration
3.5-25 mg protein/mL (protein)
50-150 nmol/mL (P450)
6-14 nmol/mg protein (specific content)
NCBI accession no.
shipped in
dry ice
Looking for similar products? Visit Product Comparison Guide
General description
Unit Definition
Other Notes
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service