Skip to Content
Merck
All Photos(1)

Documents

H3162

Supelco

Hydrochloric acid solution

1.0 N

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

concentration

1.0 N

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Jay G Hosking et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 1005-1015 (2014-10-21)
Successful decision making often requires weighing a given option's costs against its associated benefits, an ability that appears perturbed in virtually every severe mental illness. Animal models of such cost/benefit decision making overwhelmingly implicate mesolimbic dopamine in our willingness to
Xuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1365-1375 (2015-04-19)
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In
Davide R Ceratti et al.
Advanced materials (Deerfield Beach, Fla.), 27(34), 4958-4962 (2015-07-23)
A new, simple bi-phasic dip-coating method is developed. This method is considered as a great improvement of the technique for research, development and production, since expensive, rare, harmful, or time-evolving solutions can now be easily deposited on large surfaces and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service