Skip to Content
Merck
All Photos(4)

Key Documents

H0527

Sigma-Aldrich

HEPES potassium salt

HEPES potassium salt

≥99.5% (titration)

Synonym(s):

Piperazineethanesulfonic acid, 4-(2-hydroxyethyl)-monopotassium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid potassium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) potassium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$96.10
100 G
$243.00
250 G
$489.00

$96.10


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$96.10
100 G
$243.00
250 G
$489.00

About This Item

Empirical Formula (Hill Notation):
C8H17KN2O4S
CAS Number:
Molecular Weight:
276.39
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$96.10


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Quality Level

Assay

≥99.5% (titration)

form

powder

useful pH range

6.8-8.2

pKa 

7.5

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

[K+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.K/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

KWURIFKZTJGMFD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

HEPES (4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid) is a widely used Good’s buffer in biochemical and biological research applications. As a zwitterionic buffer, it stabilizes the pH of the solution[1][2] even when small amounts of acid or base are added.

Application

HEPES potassium salt has been used as a buffering agent in nanopore fabrication[3][4][5] and electrolyte preparations.[6] It has also been used as a component of cell lysis buffer.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Complexation of copper by zwitterionic aminosulfonic (good) buffers
Mash HE, et al.
Analytical Chemistry (2003)
Use of a solid-state nanopore for profiling the transferrin receptor protein and distinguishing between transferrin receptor and its ligand protein
O'Donohue M, et al.
Electrophoresis (2023)
Equilibrium studies, synthesis and characterisation of zwitterionic buffer (HEPES and HEPPS) dimethyltin (IV) complexes
Mohamed M
Main Group Chemistry, 63-84 (2007)
Determining the location of an Arabidopsis chloroplast protein using in vitro import followed by fractionation and alkaline extraction
Chu CC and Li HM
Methods in Molecular Biology (2011)
Jugal Saharia et al.
Electrophoresis, 43(5-6), 785-792 (2022-01-13)
Electrolyte chemistry plays an important role in the transport properties of analytes through nanopores. Here, we report the translocation properties of the protein human serum transferrin (hSTf) in asymmetric LiCl salt concentrations with either positive (Ctrans /Ccis < 1) or

Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    5 out of 5 stars.

    Product quality feedback

    product was good and useful as per our company requirement.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service