Skip to Content
Merck
All Photos(1)

Key Documents

H0379

Sigma-Aldrich

19-Hydroxy-4-androstene-3,17-dione

Synonym(s):

4-Androsten-19-ol-3,17-dione, 4-Androstene-3,17-dione-19-ol

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Beilstein:
2567249
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34CO)[C@@H]1CCC2=O

InChI

1S/C19H26O3/c1-18-8-7-16-14(15(18)4-5-17(18)22)3-2-12-10-13(21)6-9-19(12,16)11-20/h10,14-16,20H,2-9,11H2,1H3/t14-,15-,16-,18-,19+/m0/s1

InChI key

XGUHPTGEXRHMQQ-BGJMDTOESA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Weniger et al.
Journal of steroid biochemistry, 35(3-4), 507-510 (1990-03-01)
Ovaries from 18-21-day-old foetal as well as from 2-10-day-old infantile rats were cultured in vitro in the presence of [3H]19-hydroxyandrostenedione and in the presence or absence of LH, FSH or (Bu)2 cAMP, and oestrone and oestradiol formed were determined by
H Sekihara et al.
Clinical endocrinology, 39(5), 557-560 (1993-11-01)
We have reported that 19-hydroxyandrostenedione (19-OH-A-dione) functions as an amplifier of the sodium-retaining and hypertensive action of aldosterone. We therefore measured 19-hydroxyandrostenedione in hypertensive patients. We studied 53 normal male control subjects and 63 male patients with essential hypertension (48
K Higuchi et al.
Endocrinologia japonica, 36(6), 881-885 (1989-12-01)
The steroid, 19-hydroxyandrost-4-ene-3, 17-dione (19-hydroxyandrostene-dione, 19-OH-A-dione) has been known to enhance the mineralocorticoid action of aldosterone. To investigate the age-related change in the plasma 19-OH-A-dione concentration, plasma 19-OH-A-dione, androst-4-ene-3, 17-dione (A-dione), aldosterone and cortisol of 38 non-hypertensive healthy subjects (18
H Morita et al.
Endocrine journal, 40(1), 89-97 (1993-02-01)
The present study was performed to clarify changes in plasma levels of 19-hydroxyandrostenedione (19-OH-AD), an amplifier of aldosterone and a possible hypertensinogenic steroid, during several tests for the renin-angiotensin system in 20 patients with aldosterone-producing adenoma (APA) and to determine
S Ohigashi et al.
Nihon Naibunpi Gakkai zasshi, 65(8), 750-761 (1989-08-20)
To study the serum levels of 19-hydroxyandrostenedione (19-OH-A), known as an obligatory intermediate of estrogen biosynthesis and considered to be one of the hypertensinogens, a method using GC-MS with application of synthesized [7,7-d2]androstenedione (A), [7,7-d2] 19-OH-A and [9,11-d2]estrone(E1) as internal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service