Skip to Content
Merck
All Photos(1)

Documents

GW22654

Sigma-Aldrich

Anti-Talin-2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLN2(83660)

Immunogen

Immunogen Sequence: GI # 22035665, sequence 1750-1950
Recombinant talin 2

Application

Anti-Talin-2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Talin 2 is a protein encoded by the TLN2 gene in humans and belongs to the talin protein family. It plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. Talin 2 also plays a crutial role in cell adhesion and recycling of synaptic vesicles. It may help in serving as a link between integrins and the sarcomeric cytoskeletonin stable adhesion complexes in mature striated muscle.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa A Senetar et al.
Cell motility and the cytoskeleton, 64(3), 157-173 (2006-12-22)
The cytoskeletal protein talin serves as an essential link between integrins and the actin cytoskeleton in several similar, but functionally distinct, adhesion complexes, including focal adhesions, costameres, and intercalated disks. Vertebrates contain two talin genes, TLN1 and TLN2, but the
Lei Qi et al.
Journal of cell science, 129(19), 3661-3674 (2016-10-04)
Talin binds to β-integrin tails to activate integrins, regulating cell migration, invasion and metastasis. There are two talin genes, TLN1 and TLN2, encoding talin1 and talin2, respectively. Talin1 regulates focal adhesion dynamics, cell migration and invasion, whereas the biological function
Zheng Xiao et al.
Clinical biochemistry, 43(13-14), 1129-1132 (2010-07-14)
Talin 2 plays an important role in cell adhesion and recycling of synaptic vesicles. To explore the possible role of talin 2 in epilepsy, we designed current study to quantitatively evaluate the changes in talin 2 levels in different epilepsy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service