Skip to Content
Merck
All Photos(1)

Key Documents

GW21310

Sigma-Aldrich

Anti-RGS7 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Regulator of G-protein signaling 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-21310

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RGS7(6000)

General description

RGS7 (regulator of G-protein signaling 7) is a member of the R7 subfamily of RGS superfamily of regulators, and shares high homology with RGS9-2. It is composed of the catalytic RGS domain, a dishevelled, EGL-10 and pleckstrin (DEP) domain in its N-terminal, along with a G protein γ subunit-like (GGL) domain. It is both a soluble cytosolic protein as well as a membrane protein.

Immunogen

Immunogen Sequence: GI # 11140809, sequence 227-262
Recombinant regulator of G-protein signalling 7

Application

Anti-RGS7 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Regulator of G-protein signaling 7 belongs to a subfamily of regulator of G protein signaling (RGS) proteins, which modulate G protein signaling by acting as GTPase-activating proteins for G protein α-subunits. RGS7 exists as a dimer with the G protein β(5)-subunit. It plays a major role in the pathogenesis of panic disorder (PD). RGS7 has a potential role in neurons communicating between classical heterotrimeric G protein subunits and/or their effectors at the plasma membrane and the cell nucleus. It helps in regulating signaling by μ-opioid receptor (MOR) and dopamine D2 receptor (D2R) and are implicated in drug addiction, movement disorders and nociception.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ikuo Masuho et al.
The Journal of biological chemistry, 288(35), 25129-25142 (2013-07-17)
Regulator of G protein signaling (RGS) proteins play essential roles in the regulation of signaling via G protein-coupled receptors (GPCRs). With hundreds of GPCRs and dozens of G proteins, it is important to understand how RGS regulates selective GPCR-G protein
J H Zhang et al.
The Journal of biological chemistry, 276(13), 10284-10289 (2001-01-21)
The role that Gbeta(5) regulator of G protein signaling (RGS) complexes play in signal transduction in brain remains unknown. The subcellular localization of Gbeta(5) and RGS7 was examined in rat PC12 pheochromocytoma cells and mouse brain. Both nuclear and cytosolic
Satoshi Takida et al.
Molecular pharmacology, 67(1), 132-139 (2004-10-22)
Regulator of G protein signaling (RGS) proteins modulate G protein signaling by acting as GTPase-activating proteins for G protein alpha-subunits. RGS7 belongs to a subfamily of RGS proteins that exist as dimers with the G protein beta(5)-subunit. In this report
Christa Hohoff et al.
Journal of neural transmission (Vienna, Austria : 1996), 116(11), 1523-1528 (2008-09-03)
Following our recent finding of Rgs2 playing a role in the development of human panic disorder (PD), we examine another positional and functional candidate from the functionally interwoven Rgs (regulator of G-protein signaling) family, Rgs7, in the pathogenesis of PD.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service