Skip to Content
Merck
All Photos(1)

Key Documents

GW21035

Sigma-Aldrich

Anti-MIP-1β antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Chemokine C-C motif ligand 4 isoform 1, Anti-Macrophage inflammatory protein 1β, Anti-SCYA4, Anti-Small inducible cytokine A4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21035

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL4(6351)

General description

MIP-1β (macrophage inflammatory protein 1β) is a member of the CC chemokine (cysteine-cysteine) family, and is produced by monocytes and macrophages. This protein exists as a homodimer. It acts as a ligand to the GPCR (G-protein coupled receptor) called CCR5 (CC chemokine receptor 5).

Immunogen

Immunogen Sequence: GI # 4506845, sequence 27-92
Recombinant chemokine C-C motif ligand 4 isoform 1

Application

Anti-MIP-1β antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Chemokine (C-C motif) ligand 4 (CCL4) is a principal regulator of macrophage migration and signals through CCR5. CCR5 is linked to several protein kinases including the src kinase Lyn, PI3K, focal adhesion related kinase Pyk2 and members of the MAPK family that help in regulating macrophage chemotaxis. CCL4 is released at the site of inflammation in sarcoidosis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Barczyk et al.
International journal of immunopathology and pharmacology, 27(2), 185-193 (2014-07-10)
Sarcoidosis is a granulomatous disease with an increased accumulation of T cells in lungs as a result of on-site proliferation and chemotaxis induced by chemokines. It has already been demonstrated that CCL3-5 levels were increased in BAL fluid of sarcoidosis
Ricky Cheung et al.
Journal of leukocyte biology, 86(4), 833-845 (2009-07-22)
MIP-1beta/CCL4 is a principal regulator of macrophage migration and signals through CCR5. Several protein kinases are linked to CCR5 in macrophages including the src kinase Lyn, PI3K, focal adhesion related kinase Pyk2, and members of the MAPK family, but whether
Melissa A McCornack et al.
The Journal of biological chemistry, 278(3), 1946-1956 (2002-11-02)
Chemokines comprise a family of proteins that function in the immune response to recruit leukocytes to sites of infection. This recruitment is believed to be carried out by the establishment of a chemokine gradient by the binding of chemokines to
Isaac G Sakala et al.
Infection and immunity, 81(6), 1974-1983 (2013-03-20)
Tuberculosis remains one of the top three leading causes of morbidity and mortality worldwide, complicated by the emergence of drug-resistant Mycobacterium tuberculosis strains and high rates of HIV coinfection. It is important to develop new antimycobacterial drugs and immunomodulatory therapeutics
Li Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(38), 13985-13990 (2006-09-12)
Chemokines (chemotactic cytokines) comprise a large family of proteins that recruit and activate leukocytes, giving chemokines a major role in both immune response and inflammation-related diseases. The poxvirus-encoded viral CC chemokine inhibitor (vCCI) binds to many CC chemokines with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service