Skip to Content
Merck
All Photos(1)

Documents

GW20087F

Sigma-Aldrich

Anti-Human Kappa Light Chain antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Immunoglobulin kappa

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-20087F

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGKC(3514)

Related Categories

Immunogen

Human IgK (Kappa) native protein.

Application

Anti-Human Kappa Light Chain antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Immunoglobulin κ constant (IGKC) is a human gene that encodes the constant domain of κ-type light chains for antibodies. It is found on chromosome 2 in humans. IGKC acts as a strong prognostic marker in several human solid tumors, including breast cancer. Its expression may have a role for humoral immunity in breast cancer progression. IGKC is also found to be expressed in non-small lung cancer (NSCLC) and may act as a prognostic marker in NSCLC, supporting emerging treatment concepts that exploit the humoral immune response. Its has an independent protective impact on disease-free survival (DFS) and breast cancer-specific overall survival in node-negative breast cancer. IGKC acts as a novel diagnostic marker for risk stratification in human cancer and may help to exploit the humoral immune response for anticancer therapy.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zonglin Chen et al.
PloS one, 7(9), e44741-e44741 (2012-10-03)
Biomarkers of the immune system are currently not used as prognostic factors in breast cancer. We analyzed the association of the B cell/plasma cell marker immunoglobulin kappa C (IGKC) and survival of untreated node-negative breast cancer patients. IGKC expression was
Janardan P Pandey et al.
Immunobiology, 219(2), 113-117 (2013-09-24)
Immunoglobulin κ constant (IGKC) gene has recently been identified as a strong prognostic marker in several human solid tumors, including breast cancer. Although the mechanisms underlying the IGKC signature are not yet known, identification of tumor-infiltrating plasma cells as the
Miriam Lohr et al.
Cancer letters, 333(2), 222-228 (2013-02-02)
A prognostic impact of immunoglobulin kappa C (IGKC) expression has been described in cancer. We analysed the influence of B-cell and plasma cell markers, as well as IGKC expression, in non-small lung cancer (NSCLC) using immunohistochemistry on a tissue microarray.
Marcus Schmidt et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 18(9), 2695-2703 (2012-02-22)
Although the central role of the immune system for tumor prognosis is generally accepted, a single robust marker is not yet available. On the basis of receiver operating characteristic analyses, robust markers were identified from a 60-gene B cell-derived metagene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service