Skip to Content
Merck
All Photos(1)

Key Documents

GERPN2251

cAMP Biotrak EIA, (Non-Acetylation Protocol)

Cytiva RPN2251

Synonym(s):

cAMP Enzyme Immunoassay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116126
NACRES:
NA.32

manufacturer/tradename

Cytiva RPN2251

packaging

pkg of 96 wells

storage temp.

2-8°C

General description

EIAs and ELISAs are convenient, high-sensitivity, peroxidase-based technologies that enable the amount of protein present in a sample to be measured in a single day. Novel lysis reagents have been developed (Lysis Reagents 1 and 2) to facilitate simple and rapid extraction of cellular molecules.

The Biotrak cAMP competitive enzymeimmunoassay system is specifically designed for research purposes. The kit includes protocols using novel lysis reagents in order to facilitate simple and rapid extraction of cAMP from cell cultures. These components avoid the requirement for traditional, time-consuming extraction procedures and obviate the need for removal of extraction reagents prior to measurement. It combines the use of a peroxidase-labeled cAMP conjugate, a specific antiserum which can be immobilized on to precoated microplates, and a one-pot stabilized substrate solution.

Lysis reagent 1 hydrolyzes cell membranes to release intra-cellular
cAMP. Lysis Reagent 2 sequesters the key component in Lysis
Reagent 1 and ensures cAMP is free for subsequent analysis. The
detergent/sequestrant complex does not interfere with antigen:
antibody binding. Lysis Reagent 1 is simply added to cultured cells,
followed by a 5 to 10 min incubation before assay. The
antiserum is reconstituted with Lysis Reagent 2. The assay is based
on competition between unlabeled cAMP and a fixed quantity of
peroxidase-labeled cAMP, for a limited number of binding sites on a
cAMP specific antibody.

Application

cAMP Biotrak EIA, (Non-Acetylation Protocol) has been used to measure intracellular cyclic adenosine monophosphate (cAMP) levels.[1][2][3]

Features and Benefits

  • Elimination of inconvenient, time-consuming extraction
    procedures.
  • Flexible method-choice of assay protocols.
  • Range 25 - 6400 fmol/well (non-acetylation protocol).
  • Rapid assay protocol.
  • Nonradioactive.
  • Specific for cAMP.
  • Precise and accurate measurement
  • Ready-to-use substrate.
  • Colour coded reagents

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Biotrak is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adenosine 3′, 5′-Cyclic Monophosphate Involvement in Hepatic Triacylglyceride Lipase Release from Prazosin-Stimulated Primary Cultured Rat Hepatocytes.
Nakamura T and Morita T
Biological & Pharmaceutical Bulletin, 37(6), 922-925 (2014)
Acidosis activation of the proton-sensing GPR4 receptor stimulates vascular endothelial cell inflammatory responses revealed by transcriptome analysis.
Dong L, et al.
PLoS ONE, 8(4), e61991-e61991 (2013)
Legionella metaeffector exploits host proteasome to temporally regulate cognate effector.
Kubori T, et al.
PLoS Pathogens, 6(12), e1001216-e1001216 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service