Skip to Content
Merck
All Photos(1)

Key Documents

GEPA63101

CY®3B NHS es

Sign Into View Organizational & Contract Pricing

Select a Size

GEPA63101
$583.00

$583.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
GEPA63101
$583.00

About This Item

UNSPSC Code:
41116126
NACRES:
NA.32

$583.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

manufacturer/tradename

Cytiva

color

Red

General description

CyDye Mono-Reactive NHS Ester Fluorescent Dyes are intensely fluorescent and highly water soluble, providing significant advantages over other existing fluorophores.When labeling antibodies or other proteins with NHS esters, the optimum conditions have to be established experimentally. The extent of labeling to give maximum fluorescence between the different CyDye vary and should be taken into account it will be necessary to optimise the ratio of CyDye NHS ester to protein and pH to give the final dye to protein (D/P) ratio that is required.

Features and Benefits

  • Store refrigerated at 2–8°C in the dark.
  • Reconstitute the material to 1 mg/mL in anhydrous DMF/DMSO. Reconstituted material may be stored for up to 2 weeks at -200°C in aliquots to avoid repeat freeze thaw cycles.
  • The CyDye supplied here are monofunctional NHS esters, and are provided in a dried, pre-measured form ready for the labeling of compounds containing free amino groups.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service