GE29704234
Cytiva® UNICORN® start 1.3 control software DVD
for use with ÄKTA™ start, license activation ID
Synonym(s):
ÄKTA control software, Cytiva® control software, control software, software, DVD with license
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 29704234
compatibility
for use with ÄKTA™ start
General description
Unicorn® start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA® start instrument, and to evaluate and share results. While ÄKTA® start can function as a stand-alone system, Unicorn® start software maximizes the performance of the instrument.
Application
Features and Benefits
- Simple and flexible method creation using predefined templates and phases (steps).
- Enables real-time monitoring of the run and an intuitive way to control the run.
- Evaluate, compare, and share results. Create and print PDF reports.
- Robust database storage with easy access to data, data security, and data integrity.
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service