Skip to Content
Merck
All Photos(1)

Key Documents

GE29400461

HiTrap® Capto Core 700

Cytiva 29400461, pack of 1 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NB.13
Pricing and availability is not currently available.

packaging

pack of 1 × 1 mL

manufacturer/tradename

Cytiva 29400461

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

HiTrap Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess resin for intermediate purification and polishing of viruses and other large biomolecules. Capto Core 700 is a novel, layered bead chromatography resin consisting of a porous outer layer and ligand-activated core. The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged. These internalized ligands bind impurities strongly over a wide range of pH and salt concentration. The pores of the bead outer layer prevent large targets (Mr > 700 000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured.

Features and Benefits

  • High capacity and productivity in flowthrough mode.
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification.
  • Wide operational window of pH and conductivity.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service