Skip to Content
Merck
All Photos(1)

Key Documents

GE29360784

CyDye NIR-labelled secondary antibody

700 nm, pkg of 0.1 mg, Cytiva 29360784

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
NA.74
Pricing and availability is not currently available.

antibody product type

secondary antibodies

feature

700 nm

species reactivity

mouse

packaging

pkg of 0.1 mg

manufacturer/tradename

Cytiva 29360784

fluorescence

λex 678 nm; λem 690 nm

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Amersham CyDye NIR-labeled secondary antibodies enable quantitation of Western blots with a strong signal-to-noise ratio. Mulitplexed experiments allow for the detection of two proteins on a single blot. The use of total lane signals to normalize target protein data yield robust and reliable results.

  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation
  • Stable signals allow membranes to be stored and rescanned if needed for future studies

Amersham CyDye 700 and 800 are secondary antibodies labeled with NIR fluorophores. They emit light at wavelengths of 700 nm and 800 nm. Multiplexed experiments can be performed by using both of these secondary antibodies along with a pair of appropriate rabbit or mouse primary antibodies.

Features and Benefits

  • Easy handling and a strong signal-to-noise ratio yield reproducible results
  • Multiplexing analysis of more than one target protein on a single blot increases process efficiency
  • Total protein normalization (TPN) generates reliable and accurate quantitation
  • Stable signals allow membranes to be stored and rescanned if needed for future studies

Legal Information

CyDye is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service