GE29157185
HiTrap® MabSelect SuRe™ LX
Cytiva 29157185, pack of 5 × 5 mL
Synonym(s):
Antibody Purification Column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 29157185
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
- Reduces the risk of product contamination and carryover
- Improves performance and reduces overall costs
- High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
- Generic elution profile for different monoclonal antibodies enables platform approach to purification
- High-flow agarose matrix allows the processing of large feed volumes
Legal Information
HiTrap is a registered trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service