Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9889-38

Sigma-Aldrich

XK 16/40 Column

Synonym(s):

PCR Beads, XK Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.

material

polypropylene column

form

prepacked column

packaging

1 ea of

technique(s)

liquid chromatography (LC): suitable

General description

XK empty columns provide reliable and reproducible separations. The tube material is of borosilicate glass, which allows for visual inspection of the media bed. The columns are made of biocompatible materials that exhibit excellent chemical resistance. A broad range of column dimensions makes XK columns suitable for many different purification needs. Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations. An extensive range of spare parts is available, facilitating maintenance.

Features and Benefits

Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

Protocols

This page shows how to purify histidine-tagged proteins using TALON Superflow from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.

This page describes efficient column packing and preparation for affinity chromatography of GST-tagged proteins using Cytiva products.

This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service