Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9409-28

PD MiniTrap G-10 With Top Frit

Synonym(s):

PD MiniTrap

Sign Into View Organizational & Contract Pricing

Select a Size

GE28-9409-28
$1,550.00

$1,550.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
GE28-9409-28
$1,550.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.56

$1,550.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

manufacturer/tradename

Cytiva

General description

PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides. PD MiniTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system. PD MiniTrap G-10 columns are prepacked and designed for rapid, convenient sample clean-up of small proteins, peptides, oligo-saccharides and other small biomolecules (>700 Mr).

Application

Protein and peptide sample preparation. Desalting/Buffer Exchange/Cleanup.

Features and Benefits

  • Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700, before downstream analysis such as liquid chromatography, electrophoresis, and MS.
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels.
  • High desalting capacity.
  • Preparation of samples in the volume range 100 to 300 μl.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Legal Information

MiniTrap is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service