Skip to Content
Merck
All Photos(4)

Key Documents

GE28-9269-77

HiScreen MabSelect SuRe

Cytiva 28-9269-77

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

alkali-stabilized protein A-derived (E. coli)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9269-77

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

44 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

column I.D.

7.7 mm

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

2-13.7

working range

3-12

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

Determined at 10% breakthrough by frontal analysis at a mobile phase velocity of 500 cm/h in a column with a bed height of 20 cm.
HiScreen MabSelect SuRe is prepacked with MabSelect SuRe and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies (MAbs).
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Application

Lab Purification using the MabSelect SuRe Protein A tag for capture of antibodies.

Features and Benefits

  • The novel, alkali-tolerant recombinant protein A ligand of MabSelect Sure allows cleaning-in-place (CIP).
  • Prepacked, ready-to-use columns for convenient process development.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Free MabSelect SuRe Ligand
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
HiScreen is a trademark of Cytiva
MabSelect is a trademark of Cytiva
SuRe is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service