Skip to Content
Merck
All Photos(2)

Key Documents

GE28-9065-61

Superdex® 200 5/150 GL

Cytiva 28-9065-61, column L × I.D. 15 cm × 5 mm, 13 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

borosilicate glass column
polypropylene filter

mol wt

10000-600000 Da (globular proteins)

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 28-9065-61

parameter

0.15-0.6 mL/min flow rate
1.5 Mpa (217 psi)
4-50 mL sample volume

bed volume

3 mL

column L × I.D.

15 cm × 5 mm

matrix

dextran composite

avg. part. size

13 μm

cleaning

1-14

working range

3-12

separation technique

size exclusion (SEC)

General description

Superdex® 200 5/150 GL is a pre-packed short glass gel filtration column for rapid purity check or screening of aggregates of biomolecules in the molecular weight range 10 000 600 000, like antibodies.

Suitability

Stable in all common buffers: 1 M acetic acid, 8 M urea, 6 M guanidine hydrochloride, 30% isopropanol, 70% ethanol, 1 M NaOH (for cleaning in place)
for high-resolution, semipreperative and analytical separations of biomolecules

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Superdex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service