Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9035-31

TempliPhi Sequence Resolver Kit

Cytiva 28-9035-31, sufficient for 200 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.55
Pricing and availability is not currently available.

form

liquid

usage

sufficient for 200 reactions

feature

hotstart: no

manufacturer/tradename

Cytiva 28-9035-31

color

colorless

shipped in

dry ice

storage temp.

−70°C

General description

illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing.

The kit can be used with both small (e.g., plasmids and M13) and large (fosmids and bacterial artificial chromosomes [BACs]) circular templates. The starting template for the reaction can be purified DNA, glycerol stock, liquid culture, or colonies. The reaction takes less than 20 min to prepare and after an overnight incubation at 10° C, it is ready to be used directly in sequencing applications with the sequencing chemistry of choice. A ten microliter (10 μ) reaction typically yields 1 μg of DNA in a standard overnight reaction.

Features and Benefits

  • For the amplification of difficult templates for successful DNA sequencing.
  • Solve the most common sequencing problems such as repeats, sequencing stops, and compressions with a single kit.
  • Improve difficult template sequencing success with up to 820 bp Phred 20 read length.
  • Cultureless preparation from bacterial colonies, glycerol stock or purified DNA; saves time and reagents.
  • Easy-to-use kit requires only 20 min hands-on time.
  • Amplified DNA can be directly used in sequencing reactions.
  • illustra TempliPhi Sequence Resolver Kit produces exceptional sequencing results from difficult templates such as those with high GC content and secondary structures.
  • The kit resolves most of the sequencing stops or gaps that routinely elude other commonly used finishing methods.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
S-re the kit at -70°C. The enzyme mix must be s-red at -70°C; all other components may be s-red at -20°C Thaw components on ice and maintain at 0°C - 4°C during handling

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

TempliPhi
On packaging and insert for kits:
For use only as licensed by Qiagen GmbH. and Cytiva. The Phi 29 DNA polymerase may not be re-sold or used except in conjunction with the other components of this kit. See US patent numbers 5,854,033, 6,124,120, 6,143,495, 6,323,009, 5,198,543, 5,576,204, and their equivalents in other countries.
On container:
Subject to proprietary rights of Cytiva. May not be re-sold or used except with the other components of this kit. See US patent numbers 5,576,204, and their equivalents in other countries.
TempliPhi is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service