Skip to Content
Merck
All Photos(3)

Key Documents

GE28-4082-53

HiTrap® MabSelect

Cytiva 28-4082-53, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

recombinant protein A (E. coli)

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 28-4082-53

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

2-12

working range

3-10

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

Determined at 10% breakthrough by frontal analysis at a mobile phase velocity of 500 cm/h in a column with a bed height of 20 cm.
HiTrap® MabSelect columns are prepacked with MabSelect, a Bioprocess medium for capuring of Mabs from large sample volumes.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Application

Lab Purification using the MabSelect Protein A tag for capture of antibodies.

Features and Benefits

  • HiTrap® columns prepacked with MabSelect, a Bioprocess medium for capuring of Mabs from large sample volumes. • The specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification.
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity.
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
MabSelect is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.

This page shows how to convert between linear flow and volumetric flow rates in affinity chromatography.

How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service