Skip to Content
Merck
All Photos(1)

Key Documents

GE28-4064-18

Sigma-Aldrich

Tricorn 10/300 Column

Synonym(s):

Tricorn column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
SB.52
Pricing and availability is not currently available.

technique(s)

LPLC: suitable

General description

Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Tricorn columns include a glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock, and filter holder.

Features and Benefits

Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.

Protocols

This page covers the use of Sepharose High Performance media for purification of proteins, peptides or oligonucleotides, when to use them, and with which systems.

Column Packing and Preparation for Size Exclusion Chromatography

This page covers the use of Sepharose Fast Flow for purification of proteins.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service