Skip to Content
Merck
All Photos(1)

Key Documents

GE28-4055-00

GST MultiTrap 4B

Cytiva 28-4055-00

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.32
Pricing and availability is not currently available.

ligand

glutathione

material

polypropylene and polyethylene

manufacturer/tradename

Cytiva 28-4055-00

packaging

pkg of 4 x 96-well

size

127.8 mm × 85.5 mm × 30.6 mm

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

4-13

working range

4-13

capacity

>25 mg binding capacity (horse liver GST/ml medium)

Related Categories

General description

Binding of GST to glutathione is flow-rate dependent and lower flow rates often increase the binding capacity. This is important to consider during sample loading and elution.
GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and rapid parallel purification of glutathione S-transferase (GST) tagged proteins.

Application

GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and purification of glutathione S-transferase (GST) tagged proteins.

Purification of up to 0.5 mg of GST-tagged Proteins/well directly from unclarified cell lysate can be achieved using GST MultiTrap filter plates. Direct purification of GST-tagged Proteins from unclarified lysate shortens handling time and minimizes degradation of sensitive target Proteins. The 96-well plate format gives great flexibility, both when working with automated robotic systems and manually using centrifugation or vacuum. Consistent well-to-well and plate-to-plate performance ensures high reproducibility.

GST MultiTrap FF is prepacked with Glutathione Sepharose 4 Fast Flow, while GST MultiTrap 4B is prepacked with Glutathione Sepharose 4B.

Features and Benefits

  • 96-well filter plate format for high-throughput screening and parallel purification of GST-tagged* Proteins.
  • Highly reproducible well-to well and plate-to-plate results.
  • High purity in one step, high chemical stability. Load unclarified samples directly to the wells.
  • Wells are prepacked with Glutathione Sepharose media for convenience.
  • Easy and predictable scale-up using prepacked GSTrap FF, GSTPrep FF 16/10, and GSTrap 4B columns.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

GSTPrep is a trademark of Cytiva
GSTrap is a trademark of Cytiva
MultiTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows troubleshooting methods for purification of GST-tagged proteins using Cytiva products.

This page describes characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (high sub) media and columns from Cytiva.

This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.

Protocols

This page how to remove GST tags by enzymatic cleavage using Cytiva products.

This page shows how to solubilize membrane proteins with products from Cytiva.

Glutathione Sepharose® High Performance is recommended for high-resolution purification of GST-tagged proteins, providing sharp peaks and concentrated eluent.

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service