Skip to Content
Merck
All Photos(1)

Documents

GE27-9606-02

Cyscribe GFX Purification Kit

Cytiva 27-9606-02, pack of 50 columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

packaging

pack of 50 columns

manufacturer/tradename

Cytiva 27-9606-02

General description

illustra CyScribe GFX Purification Kit has been specifically developed for the purification of CyDye labeled cDNA probes. Allows rapid purification of CyDye labeled cDNA probes.

Application

illustra CyScribe GFX Purification Kit has been specifically developed for the purification of CyDye labeled cDNA probes. It can be used in conjunction with CyScribe microarray labeling kits for either direct incorporation of Cy3- and Cy5-labeled dNTPs or for post-labeling of cDNA with Cy3 and Cy5 monoreactive dyes.

Features and Benefits

  • Provides efficient removal of unincorporated CyDye label and primers from labeling reactions with outstanding yields of labeled cDNA probe.
  • Excellent for the purification of cDNA labeled by either direct incorporation or by post-labeling using CyScribe labeling kits.
  • Optimized purification reagents are conveniently available in both 25 and 50 column format or can be purchased with the CyScribe labeling kits.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Cy3 is a trademark of Cytiva
CyDye is a trademark of Cytiva
GFX is a trademark of Cytiva
illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

"" - ""


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service