Skip to Content
Merck
All Photos(1)

Key Documents

GE17548801

Cytodex® 3 Gamma Microcarriers

sterile, dry powder, Cytiva, pkg of 30 g

Sign Into View Organizational & Contract Pricing

Select a Size

30 G
$2,350.00

$2,350.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
30 G
$2,350.00

About This Item

UNSPSC Code:
41161502
NACRES:
NC.01

$2,350.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

sterility

sterile

feature

dry powder

packaging

pkg of 30 g

manufacturer/tradename

17548801
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.

  • Gamma-sterilized and ready to use for quick culture start-up.
  • Delivered in a convenient, single-use container system for easy transfer to various cell culture vessels.
  • Supplied dry to save storage space and facilitate transportation.
  • Enable culturing of anchorage-dependent cells in single-use bioreactor to eliminate the need for cleaning and cleaning validation.
  • Supported by extensive product documentation to facilitate regulatory filing.
  • Serum-free function testing to ensure performance.


Successful serial subcultivation protocols
Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix. As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity. This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service