Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5458-11

HiTrap® 5x1 ml, KappaSelect

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,560.00

$1,560.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.


Select a Size

Change View
5 ML
$1,560.00

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

$1,560.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

manufacturer/tradename

Cytiva

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked agarose

average diameter

75 μm

cleaning

2-10

working range

3-10

suitability

suitable for bioprocess medium

General description

  • KappaSelect is an affinity chromatography resin designed for the purification of human Fab (kappa) fragments.
  • It is a rigid agarose base matrix that facilitates high flow rates and large volume processing of samples for efficient throughput.
  • The Fab purity and productivity are maintained by the low ligand leakage.
  • Non-mammalian derived product is also effective in binding IgG, IgA, and IgM target molecules.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 8 °C. (20% Ethanol)

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service