Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5195-01

HiTrap® Phenyl High Performance

Cytiva 17-5195-01, pack of 5 × 5 mL

Synonym(s):

HiTrap Phenyl Column, Phenyl High Performance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

phenyl

packaging

pack of 5 × 5 mL

manufacturer/tradename

Cytiva 17-5195-01

parameter

42 psi

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning

2-14

working range

3-13

suitability

suitable for bioprocess medium (BioProcess medium.)
suitable for bioprocess medium

Related Categories

General description

HiTrap® Phenyl HP are prepacked Phenyl Sepharose High Performance columns for high-resolution hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.

Features and Benefits

  • Provide convenient and fast small-scale protein purifications.
  • Phenyl Sepharose High Performance is a well-established aromatic HIC medium, designed for intermediate and polishing steps where high resolution has priority.
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as AKTA design.
  • High flow rates and high capacities.
  • Excellent and easy scale-up achieved by connecting several HiTrap® columns in series or using HiPrep columns.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiPrep is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

This page discusses column packing and preparation techniques for reverse phase chromatography.

Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.

Protocols

Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.

This page shows how to condition membrane proteins for further analysis with products from Cytiva.

This page discusses various aspects of sample preparation for chromatographic purification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service