Skip to Content
Merck
All Photos(2)

Key Documents

GE17-5130-02

GSTrap Fast Flow

Cytiva 17-5130-02, pack of 2 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.25
Pricing and availability is not currently available.

ligand

glutathione and 10-carbon linker arm

packaging

pack of 2 × 1 mL

manufacturer/tradename

Cytiva 17-5130-02

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning

3-12

working range

3-12

capacity

>10 mg/mL binding capacity

General description

GSTrap FF columns are prepacked Glutathione Sepharose Fast FLow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Application

GST*-tagged Proteins can be purified directly from pretreated bacterial lysates using GSTrapTM FF.GST-tagged Proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves Protein antigenicity and function. If desired, cleavage of the Protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged Proteins can be detected using colorimetric or immunological methods. The medium, Glutathione SepharoseTM 4 Fast Flow, is also available as lab packages and is an excellent choice for scale-up.The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTATM design or FPLCTM System.

Features and Benefits

  • Fast and simple one-step purification of GST-tagged Proteins.
  • Mild elution conditions preserving Protein antigenicity and function.
  • Easy scale-up by connecting the columns in series.
  • Sepharose 4 Fast FLow provides good fLow properties.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

GSTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows troubleshooting strategies for cleavage methods using Cytiva products.

This page shows how to cleave and purify GST-tagged proteins eluted from GSTrap from Cytiva.

This page shows how to cleave and purify GST-tagged proteins bound to GSTrap from Cytiva.

This page shows troubleshooting methods for purification of GST-tagged proteins using Cytiva products.

Protocols

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

GSTrap™ affinity columns are specially designed 1 ml and 5 ml HiTrap® columns packed with Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, or Glutathione Sepharose 4B media.

This page how to remove GST tags by enzymatic cleavage using Cytiva products.

Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service