Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5068-01

Sigma-Aldrich

SOURCE 15RPC ST 4.6/100

pkg of 1 column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.

ligand

matrix surface

material

polypropylene column

feature

autoclavable 20 min at 121°C

packaging

pkg of 1 column

parameter

flow rate
0.5 ml/min - 2.5 ml/min flow rate
580 psi

technique(s)

HPLC: suitable

bed size

4.6 mm × 100 mm

bed volume

1.7 mL

column I.D.

4.6 mm

matrix

polymeric
polystyrene/divinylbenzene

matrix active group

polystyrene-divinylbenzene phase

avg. part. size

15 μm

cleaning

1-14

working range

1-12

capacity

~10 mg binding capacity

suitability

suitable for bioprocess medium

separation technique

reversed phase

General description

SOURCE 15RPC ST 4.6/100 is a prepacked stainless steel column which provides fast, high-capacity preparative protein purification with reversed phase chromatography.
~10 mg BSA /ml medium at 300 cm/h, ~30 mg bacitracin/ml medium at 300 cm/h, ~50 mg insulin/ml medium at 300 cm/h

Application

SOURCE 15RPC is based on rigid, monosized 15 μm diameter polystyrene/divinyl benzene beads. The matrix has outstanding selectivity for RPC. The monodispersity of the beads yields stable beds, Low back pressures and excellent results at high flow rates.With its high physical and chemical stability and very high batch-to-batch reproducibility, SOURCE 15RPC is well suited for all stages of an industrial scale operation - from research and process development through scale-up and into production. SOURCE 15RPC offers properties superior to those of other polymeric matrices.

Features and Benefits

  • Wide pH range (1-12), outstanding selectivity, chemical resistance, high capacity, and high resolution at high fLow rates.
  • Excellent scalability, from RESOURCE to FineLINE columns.
  • using ÄKTA® design and other high-performance liquid chromatography systems.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol or 70% Acetonitrile)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

RESOURCE is a trademark of Cytiva
SOURCE is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

“Ghosting” in chromatograms due to poor eluent quality; trace organic impurities concentrate, causing unknown peaks.

The different stages of working with SOURCE™ (Cytiva) for high-resolution separation are discussed, from purification to polishing and scaling up

This page discusses column packing and preparation techniques for reverse phase chromatography.

Protocols

In this section the practical aspect of Reverse Phased Chromatography ( RPC) is discussed including media and column selection and eluent selection and preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service