Skip to Content
Merck
All Photos(1)

Key Documents

GE17-3719-11

HiTrap® 5x1 mL Capto Butyl ImpRes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,110.00

$1,110.00


Usually ships in 2 weeks.


Select a Size

Change View
1 EACH
$1,110.00

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

$1,110.00


Usually ships in 2 weeks.

shelf life

5 years from manufacture date.

manufacturer/tradename

Cytiva

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

high-flow agarose

suitability

suitable for bioprocess medium

Related Categories

General description

HiTrap Capto Butyl ImpRes is a ready-to-use column prepacked with BioProcess Capto Butyl ImpRes media for screening and small-scale protein purification using hydrophobic interaction chromatography.

  • Prepacked columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
  • High-throughput chromatography medium for intermediate and polishing steps in a downstream protein purification process.
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide.
  • High binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.


Features and Benefits

  • Prepacked columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
  • High-throughput chromatography medium for intermediate and polishing steps in a downstream protein purification process.
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide.
  • High binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service