Skip to Content
Merck
All Photos(7)

Key Documents

GE17-3717-02

Capto S ImpAct

Cytiva 17-3717-02, pack of 100 ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,000.00

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EACH
$1,000.00

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

ligand

(-SO3)

manufacturer/tradename

Cytiva 17-3717-02

matrix

high-flow agarose

avg. part. size

50 μm (d50V)

cleaning in place

4-12

working range

3-14

capacity

binding capacity(>85 mg BSA/mL medium, >90 mg lysozyme/ml medium, >100 mg IgG/ ml medium)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This chromatography medium is part of our platform of high-resolution media based on the Capto product line. The polymeric ligand, in combination with the attributes of the base matrix, gives this cation exchanger a high binding capacity. Furthermore, the small bead size enables high-resolution purifications.

The high-flow characteristics of Capto media allows for excellent pressure-flow properties. The exceptional binding capacity and resolution of Capto S ImpAct, combined with the ability to run at high flow rates and bed heights, increase productivity and flexibility in process design.

Capto S ImpAct is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development.

Capto S ImpAct is available in a range of different bulk pack sizes and convenient prepacked formats for easy scale-up and process development. As a member of the BioProcess media range, Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

Capto S ImpAct is a strong cation exchange chromatography medium (resin) developed for intermediate purification and polishing of monoclonal antibodies (MAbs) and other biomolecules.

  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution intermediate purification and polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
  • High productivity enabling cost-efficient manufacturing.
  • Security of supply and comprehensive regulatory support.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service