Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1180-01

Resource S

Cytiva 17-1180-01, pack of 6 mL

Synonym(s):

Resource S Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

sulfonate group

description

Ion Exchanger Type (value)

packaging

pack of 6 mL

manufacturer/tradename

Cytiva 17-1180-01

parameter

87 psi

bed size

16 mm × 30 mm

bed volume

6 mL

matrix

polystyrene/divinylbenzene

avg. part. size

15 μm

particle size

15-15 μm

cleaning

1-14

working range

2-13

suitability

suitable for bioprocess medium (BioProcess medium. Columns not suitable for use with ÄKTAprime plus system.)
suitable for bioprocess medium

General description

RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.

The small (15 μm) monodisperse bead give high-resolution purification at high flow rates. In addition, hydrophilization of the beads minimizes nonspecific adsorption and allows high recovery of purified sample. The material of the column body is PEEK (polyetheretherketone). SOURCE 30Q and 30S media with a 30 μm bead size are designed for intermediate purification and large-scale polishing and allow higher flow rates.

Features and Benefits

  • SOURCE 15Q and 15S media are well suited for fast, high-resolution purifications and easy scale-up.
  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25 mg of protein/mL, and improved stability in organic solvents and pH extremes
  • RESOURCE prepacked columns for fast separation. Separation times using 1 mL RESOURCE columns are less than 3 min at 9.6 mL/min and about 20 min using a peristaltic pump at 1 mL/min.
  • Improved capacity compared to MonoBeads but with a slightly lower resolution.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

RESOURCE is a trademark of Cytiva
SOURCE is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to perform a purification of His-tagged membrane proteins.

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

See All

Protocols

This page covers using SOURCE™ medias for the purification of proteins, peptides, or oligonucleotides.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service