Skip to Content
Merck
All Photos(1)

Documents

GE17-1128-01

MAbtrap® Kit

Cytiva 17-1128-01, pkg of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

recombinant protein G lacking albumin-binding region

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva 17-1128-01

storage condition

(20% Ethanol)

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning

2-9

working range

3-9

capacity

>1 mg binding capacity (lgG/column)

storage temp.

2-8°C

General description

MAbTrap® is an affinity chromatography kit, with everything required for small-scale IgG purification from ascites fluid, serum, and cell culture supernatants.
The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Application

MAbTrap® Kit is an affinity chromatography kit for fast and effective purification of monoclonal and polyclonal IgG from ascites fluid, serum, and cell culture supernatants. MAbTrap® Kit contains everything necessary to perform a complete one step purification without using complicated equipment. It also eliminates time-consuming buffer preparation and column packing. The kit includes one HiTrap® column, prepacked with Protein G Sepharose High Performance.

Features and Benefits

  • Convenient kit for purification of IgG from monoclonal and polyclonal material using a syringe in less than 15 min.
  • Kit contains one HiTrap® Protein G HP column prepacked with Protein G Sepharose High Performance and enough buffer concentrate for 20 preparations with a syringe.
  • MAbTrap® Kit contains everything necessary to perform a complete one-step purification without using complex equipment.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
MAbTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Purify monoclonal or polyclonal IgG from serum, cell culture supernatant or ascitic fluid using the HiTrap Protein G HP from Cytiva, an affinity-exclusion chromatography product containing Sepharose-immobilized Protein G.

This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service