Skip to Content
Merck
All Photos(2)

Key Documents

GBL805020

Grace Bio-Labs PATH® protein microarray slides, with barcode

nitrocellulose (on glass slide), external size 25 mm × 75 mm, pack of 20 ea

Synonym(s):

barcoded non-porous nitrocellulose film slide

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$1,270.00

$1,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
20 EA
$1,270.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.15

$1,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

nitrocellulose (on glass slide)

packaging

pack of 20 ea

manufacturer/tradename

Grace Bio-Labs 805020

external size

25 mm × 75 mm

General description

The Grace Bio-Labs PATH protein microarray slides are a non-porous nitrocellulose film slides.

Benefits:
  • Low and consistant fluorescent backgrounds
  • Higher protein binding
  • Superior signal-to-noise

Application

  • Epitope-mapping
  • Biomarker Discovery and Validation
  • Coronavirus proteome microarray
  • Protein glycosylation profiling
  • Antibody Validation
  • Matrix-Assisted Laser desorption/Ionization Mass Spectrometry Imaging (MALDI MSI)

Legal Information

PATH is a registered trademark of Courtagen Life Sciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service