Skip to Content
Merck
All Photos(2)

Key Documents

GBL705024

Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides

24-5 mm x 6 mm pads per slide, plain glass, barcoded

Synonym(s):

nitrocellulose slides, protein microarrays

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$1,150.00

$1,150.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
20 EA
$1,150.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

$1,150.00


Estimated to ship onMay 29, 2025


material

nitrocellulose (on glass)

packaging

pkg of 20 ea

manufacturer/tradename

Grace Bio-Labs 705024

external size

25 mm × 75 mm

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Porous nitrocellulose (PNC) film slides are the major high binding protein microarray substrate technology. Grace Bio-Labs ONCYTE® nitrocellulose film slides offer a significantly higher surface area for protein binding. These slides are compatible with many detection methods. Fluorescent, chemiluminescent, isotopic, and chromogenic detection methods that are commonly employed for Western and Northern blots are compatible with porous nitrocellulose films. ONCYTE® film slides need not require any activation step to immobilize proteins and are printable straight from the box. Pre-wetting ONCYTE® film slides should be avoided as printing onto a wet slide will result in the sample spreading, resulting in larger, more diffuse spots.

Application

Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides might be suitable for:
  • antigen or antibody capture
  • cell or tissue lysate binding
  • glycoprotein and peptide binding assay

Features and Benefits

  • Packed in boxes with minimal off-gassing
  • Require no pre-processing prior to array spotting
  • Ready for printing straight from the box
  • Require no activation steps to immobilize proteins
  • Can be analyzed using various laser scanners or colorimetric or fluorescence detectors (CCD) imaging systems

Legal Information

NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service