Skip to Content
Merck
All Photos(2)

Key Documents

GBL664112

Grace Bio-Labs FastWells reagent barriers

9 mm diam. x 1.0 mm depth

Synonym(s):

flexible silicone slide gasket, reagent chamber

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$228.00

$228.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$228.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

$228.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

feature

wells 1

packaging

pack of 50 ea

manufacturer/tradename

Grace Bio-Labs 664112

diam. × depth

9 mm × 1.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

FastWells reagent barriers are sticky, flexible silicone gaskets that form hydrophobic barriers around specimens without messy adhesives or special slides. Gaskets may be stacked to increase depth and volume.

FastWells provide rapid isolation of cells and tissues on slides or coverslips for antibody incubations. The barriers remain sealed during agitated washing steps, preventing reagents from spilling when shaken or rocked. FastWells peel off quickly and cleanly, leaving no residue to interfere with specimen cover-slipping. They can be washed, autoclaved and reused. The barriers can also be sealed to form incubation chambers using flexible HybriSlip covers.

Legal Information

FastWells is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service