GBL620005
Grace Bio-Labs SecureSeal™ adhesive sheets
thickness 0.60 mm, Double Sided, L × W 7 in. × 10 in.
Synonym(s):
silicone sheet with double sided adhesive
Select a Size
$285.00
Select a Size
About This Item
$285.00
Recommended Products
sterility
non-sterile
feature
Double Sided
packaging
pack of 1 ea
manufacturer/tradename
Grace Bio-Labs 620005
L × W
7 in. × 10 in.
thickness
0.60 mm
Related Categories
General description
Grace Bio-Labs SecureSeal™ adhesive sheets are made up of SecureSeal™ adhesive, which is used to make HybriWell™ and SecureSeal™ incubation chambers. It has a thin, double-sided tape on a non-woven carrier, a clear, polyester liner on one surface, and a white clay-coated paper on the other. SecureSeal™ sheets are available in various thicknesses.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)