GBL612102
Grace Bio-Labs HybriWell™ sealing system, SecureSeal™ adhesive chamber
chamber volume 280-330 μL, L × W × thickness 22 mm × 60 mm × 0.25 mm, port diam. 1.5 mm
Synonym(s):
chamber slides, chambered coverglass, cover slides, hybridization chambers, incubation chambers
Select a Size
$557.00
Select a Size
About This Item
$557.00
Recommended Products
material
colorless
polycarbonate
feature
wells 1
packaging
pack of 100
manufacturer/tradename
Grace Bio-Labs 612102
L × W × thickness
22 mm × 60 mm × 0.25 mm
chamber volume
280-330 μL
port diam.
1.5 mm
wells
1
Looking for similar products? Visit Product Comparison Guide
General description
Features:
- Encloses a single or multiple specimens in a small reagent volume and eliminate evaporation
- Access ports in the chamber surface allow for the addition or removal of reactants
- Ports are easily sealed using adhesive seal tabs provided with each order
- Sealed chambers are watertight and ideally suited for waterbath incubations
- RNase free, hydrophobic surfaces will not trap or bind probes
- Disposable chamber removes cleanly and easily even after heating
- Each package includes illustrated instructions for use, applicator and 200 adhesive port seals
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service