GBL248864
Grace Bio-Labs ProPlate® microarray system, slide module
W × L 16 mm × 16 mm, with stainless steel spring clips, wells, 4
Synonym(s):
incubation chambers, microarray hybridization chambers, slide chambers
Select a Size
$136.00
Select a Size
About This Item
$136.00
Recommended Products
material
black silicon
square wells
sterility
non-sterile
feature
lid: no
wells 4
packaging
pack of 2 ea
manufacturer/tradename
Grace Bio-Labs 248864
W × L
16 mm × 16 mm
size
4 wells
well volume
350 μL
color
none
Looking for similar products? Visit Product Comparison Guide
General description
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
- Up to 64 leak-proof wells on any slide surface without the use of adhesive
- Microtiter plate well spacing facilitates use with multichannel pipettes
- 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service