Skip to Content
Merck
All Photos(1)

Key Documents

G9785

Sigma-Aldrich

GSEM Supplement

liquid, suitable for cell culture,

Synonym(s):

Glutamine synthetase expression medium supplement

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$165.00

$165.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$165.00

About This Item

MDL number:
UNSPSC Code:
12352205
NACRES:
NA.75

$165.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

GSEM Supplement, 50 ×, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

Quality Level

form

liquid

concentration

50 ×

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Glutamine synthetase expression medium (GSEM) supplement enables formation of glutamine from ammonia and glutamate there by alleviating accumulation of ammonia in culture medium.[1] It is exploited for high yield recombinant protein production in Chinese hamster ovary (CHO) cells.[2]

Application

GSEM Supplement has been used as a supplement for ProCHO5 Protein-free medium for culturing Chinese hamster ovary (CHO) cells[3] and selective medium for NS0 myeloma cells.[4]

Packaging

Contains (mg/L): L-alanine 450.0, L-asparagine · H2O 4261.0, L-aspartic acid 650.0, L-glutamic acid 3750.0, L-proline 575.0, L-serine 500.0, adenosine 350.0, guanosine 350.0, cytidine 350.0, uridine 350.0, and thymidine 12.0

Quantity

One mL will supplement 50 mL of cell culture medium.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble Fc fusion proteins for biomedical research
Monoclonal Antibodies: Methods and Protocols, 33-52 (2007)
Comprehensive characterization of glutamine synthetase-mediated selection for the establishment of recombinant CHO cells producing monoclonal antibodies
Noh SM, et al.
Scientific reports, 8(1), 5361-5361 (2018)
A streamlined implementation of the glutamine synthetase-based protein expression system
Knox R, et al.
BMC biotechnology, 13(1), 74-74 (2013)
Novel membrane?bound reporter molecule for sorting high producer cells by flow cytometry.
Helman D, et al.
Cytometry. Part A : the Journal of the International Society For Analytical Cytology, 85(2), 162-168 (2014)
Doyle, A., et al.
Cell and Tissue Culture: Laboratory Procedures, 27D, 3-3 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service