Skip to Content
Merck
All Photos(1)

Key Documents

G9416

Sigma-Aldrich

Glucagon-Like Peptide 1 Fragment 7-37 human

≥96% (HPLC)

Synonym(s):

GLP-1, 7-37, Preproglucagon 78-108

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$708.00

$708.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$708.00

About This Item

Empirical Formula (Hill Notation):
C151H228N40O47
Molecular Weight:
3355.67
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

$708.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥96% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)Cc4c[nH]cn4)[C@@H](C)O)[C@@H](C)O)C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(O)=O

InChI

1S/C151H228N40O47/c1-17-77(10)121(148(236)169-81(14)127(215)177-105(60-87-63-160-92-36-25-24-35-90(87)92)138(226)179-101(56-74(4)5)139(227)188-119(75(6)7)146(234)176-94(37-26-28-52-152)130(218)161-65-111(199)170-93(39-30-54-159-151(156)157)129(217)164-68-118(210)211)190-140(228)103(57-84-31-20-18-21-32-84)180-135(223)99(47-51-116(206)207)175-134(222)95(38-27-29-53-153)172-125(213)79(12)166-124(212)78(11)168-133(221)98(44-48-110(155)198)171-112(200)66-162-132(220)97(46-50-115(204)205)174-136(224)100(55-73(2)3)178-137(225)102(59-86-40-42-89(197)43-41-86)181-143(231)107(69-192)184-145(233)109(71-194)185-147(235)120(76(8)9)189-142(230)106(62-117(208)209)182-144(232)108(70-193)186-150(238)123(83(16)196)191-141(229)104(58-85-33-22-19-23-34-85)183-149(237)122(82(15)195)187-113(201)67-163-131(219)96(45-49-114(202)203)173-126(214)80(13)167-128(216)91(154)61-88-64-158-72-165-88/h18-25,31-36,40-43,63-64,72-83,91,93-109,119-123,160,192-197H,17,26-30,37-39,44-62,65-71,152-154H2,1-16H3,(H2,155,198)(H,158,165)(H,161,218)(H,162,220)(H,163,219)(H,164,217)(H,166,212)(H,167,216)(H,168,221)(H,169,236)(H,170,199)(H,171,200)(H,172,213)(H,173,214)(H,174,224)(H,175,222)(H,176,234)(H,177,215)(H,178,225)(H,179,226)(H,180,223)(H,181,231)(H,182,232)(H,183,237)(H,184,233)(H,185,235)(H,186,238)(H,187,201)(H,188,227)(H,189,230)(H,190,228)(H,191,229)(H,202,203)(H,204,205)(H,206,207)(H,208,209)(H,210,211)(H4,156,157,159)/t77-,78-,79-,80-,81-,82+,83+,91-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,119-,120-,121-,122-,123-/m0/s1

InChI key

GCYXWQUSHADNBF-AAEALURTSA-N

Gene Information

human ... GCG(2641)

Amino Acid Sequence

His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala- Trp-Leu-Val-Lys-Gly-Arg-Gly

General description

GLP-1 (7-37) is a truncated, bioactive form of GLP-1 that is the product of proglucagon processing in intestinal endocrine L cells. It is a potent insulinotropic hormone.
Glucagon-Like Peptide 1 Fragment 7-37 human (GLP-1-(7-37)) is an amino-truncated form of GLP-1.[1]

Application

Glucagon-Like Peptide 1 Fragment 7-37 human has been used:
  • as a positive control for insulin secretion of pancreatic islets[2]
  • to test its effect on insulin response in horse islets[3]
  • in combination with mesenchymal stem cells (MSCs) test its protective effects in myocardial infarction[4]

Biochem/physiol Actions

Glucagon-Like Peptide 1 Fragment 7-37 human (GLP-1-(7-37)) interacts with the GLP-1 receptor (GLP-1r). It mediates insulin release[5] by stimulating the cyclic AMP accumulation in islet cells [1] GLP-l(7-37) is a potential therapeutic agent due to its insulin-secretagogue functionality.[6]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth J Wright et al.
Stem cells international, 2016, 7357096-7357096 (2016-12-23)
Background. Mesenchymal stem cells (MSCs) and glucagon-like peptide-1 (GLP-1) are being tested as treatment strategies for myocardial infarction (MI); however, their mechanisms in the heart are not fully understood. Methods. We examined the effects of MSCs, either native, or engineered
D M Hargrove et al.
Metabolism: clinical and experimental, 44(9), 1231-1237 (1995-09-01)
In vitro, truncated glucagon-like peptides [GLP-1(7-36)-amide and GLP-1(7-37)] increase insulin secretion in a glucose-dependent manner, and desensitization to the action of GLP-1(7-37) has been demonstrated acutely with high concentrations. The purpose of these studies was to evaluate the glucose dependency
Torsten P Vahl et al.
The Journal of clinical endocrinology and metabolism, 88(4), 1772-1779 (2003-04-08)
Glucagon-like peptide 1 (GLP-1) is an insulin secretagogue synthesized in the intestine and released in response to meal ingestion. It is secreted primarily in two forms, GLP-1-(7-37) and GLP-1-(7-36)NH(2), both of which bind to a specific GLP-1 receptor (GLP-1r) on
Daniel J Drucker et al.
The Journal of clinical investigation, 127(12), 4217-4227 (2017-12-05)
The discovery, characterization, and clinical development of glucagon-like-peptide-1 (GLP-1) spans more than 30 years and includes contributions from multiple investigators, science recognized by the 2017 Harrington Award Prize for Innovation in Medicine. Herein, we provide perspectives on the historical events
Murad H Kheder et al.
PeerJ, 6, e4316-e4316 (2018-02-07)
Equine metabolic syndrome (EMS) is associated with insulin dysregulation, which often manifests as post-prandial hyperinsulinemia. Circulating concentrations of the incretin hormone, glucagon-like peptide-1 (GLP-1) correlate with an increased insulin response to carbohydrate intake in animals with EMS. However, little is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service