Skip to Content
Merck
All Photos(1)

Key Documents

G9031

Sigma-Aldrich

Gly-Val-Leu-Ser-Asn-Val-Ile-Gly-Tyr-Leu-Lys-Lys-Leu-Gly-Thr-Gly-Ala-Leu-Asn-Ala-Val-Leu-Lys-Gln trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

PDQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H194N30O31
CAS Number:
Molecular Weight:
2456.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)CN)C(C)C)C(C)C)C(=O)NCC(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C112H194N30O31.C2HF3O2/c1-22-62(18)91(142-111(171)90(61(16)17)141-105(165)79(48-83(119)148)134-106(166)80(53-143)137-103(163)76(45-58(10)11)135-109(169)88(59(12)13)138-84(149)49-116)107(167)121-51-86(151)125-77(46-66-32-34-67(145)35-33-66)104(164)132-74(43-56(6)7)100(160)127-68(29-23-26-38-113)96(156)126-70(31-25-28-40-115)98(158)131-72(41-54(2)3)95(155)120-52-87(152)139-92(65(21)144)108(168)122-50-85(150)123-63(19)93(153)130-73(42-55(4)5)102(162)133-78(47-82(118)147)99(159)124-64(20)94(154)140-89(60(14)15)110(170)136-75(44-57(8)9)101(161)128-69(30-24-27-39-114)97(157)129-71(112(172)173)36-37-81(117)146;3-2(4,5)1(6)7/h32-35,54-65,68-80,88-92,143-145H,22-31,36-53,113-116H2,1-21H3,(H2,117,146)(H2,118,147)(H2,119,148)(H,120,155)(H,121,167)(H,122,168)(H,123,150)(H,124,159)(H,125,151)(H,126,156)(H,127,160)(H,128,161)(H,129,157)(H,130,153)(H,131,158)(H,132,164)(H,133,162)(H,134,166)(H,135,169)(H,136,170)(H,137,163)(H,138,149)(H,139,152)(H,140,154)(H,141,165)(H,142,171)(H,172,173);(H,6,7)/t62-,63-,64-,65+,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,88-,89-,90-,91-,92-;/m0./s1

InChI key

BURHHWGBXFIPSQ-FAJNZGOOSA-N

Biochem/physiol Actions

Antimicrobial peptide originally isolated from Xenopus laevis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucinda K Bell et al.
Eating behaviors, 31, 28-34 (2018-08-08)
Previous studies have investigated associations between individual foods or food group intake, and breastfeeding duration, age of solid introduction and food neophobia. This study aimed to investigate associations between whole dietary patterns in young children, and breastfeeding duration, age of
Andrea Burri et al.
International journal of impotence research, 30(4), 171-178 (2018-05-26)
The aim of the present study was to investigate which PD specific factors (e.g., degree of penile curvature, levels of pain) cause most distress and to further explore whether there are specific subgroups of patients that report particularly high levels
Christine H Meyer-Frießem et al.
The Clinical journal of pain, 35(2), 111-120 (2018-09-28)
Spinal cord and peripheral nerve stimulation (SCS/PNS) may alleviate chronic pain; however, the underlying mechanisms remain controversial. The aim of this observational study was to assess sensory changes in the ON-conditions and OFF-conditions to obtain insights into the mechanism of
Kapil Gudala et al.
Pain practice : the official journal of World Institute of Pain, 17(8), 1042-1049 (2017-02-06)
The contribution of neuropathic pain (NeP) in chronic pain conditions is increasingly emphasized with the development of multiple questionnaire-based measurement scales. The painDETECT questionnaire (PDQ) is a frequently used self-reported outcome measure to assess NeP in patients with chronic pain
Rupert McShane et al.
The Cochrane database of systematic reviews, 3, CD003154-CD003154 (2019-03-21)
Memantine is a moderate affinity uncompetitive antagonist of glutamate NMDA receptors. It is licensed for use in moderate and severe Alzheimer's disease (AD); in the USA, it is also widely used off-label for mild AD. To determine efficacy and safety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service