Skip to Content
Merck
All Photos(1)

Key Documents

G8296

Sigma-Aldrich

GRK5, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GPRK5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

32-44 nmol/min·mg

mol wt

~95 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GRK5(2869)

General description

GRK5 (G protein-coupled receptor kinase 5) belongs to the G-protein coupled receptor kinase family, and shows major expression in normal human heart. This protein contains a nuclear localization sequence and hence localizes in nucleus.[1][2]

Biochem/physiol Actions

GRK5 (G protein-coupled receptor kinase 5) controls the functions of catecholamines, which bind and activate β-adrenergic receptors, and have cardiac inotropic and chronotropic actions. It is responsible for the uncoupling from G protein by facilitating β1-adrenergic receptor phosphorylation and β-arrestin recruitment.[1] This protein plays a key role in growth and metastasis in prostate cancer, where it phosphorylates moesin.† A genome-wide association study (GWAS) has shown that rs10886471 variant in GRK5 gene is linked with susceptibility to type 2 diabetes mellitus (T2DM).[3]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prabir Kumar Chakraborty et al.
Cancer research, 74(13), 3489-3500 (2014-04-24)
G protein-coupled receptor kinases (GRK) regulate diverse cellular functions ranging from metabolism to growth and locomotion. Here, we report an important contributory role for GRK5 in human prostate cancer. Inhibition of GRK5 kinase activity attenuated the migration and invasion of
Miklos D Kertai et al.
Circulation. Cardiovascular genetics, 7(5), 625-633 (2014-07-23)
We hypothesized that genetic variations in the adrenergic signaling pathway and cytochrome P450 2D6 enzyme are associated with new-onset atrial fibrillation (AF) in patients who underwent coronary artery bypass grafting and were treated with perioperative β-blockers (BBs). Two cohorts of
Zhenfang Xia et al.
PloS one, 9(3), e90597-e90597 (2014-03-07)
A genome-wide association study had showed G-protein-coupled receptor kinase 5 (GRK5) rs10886471 was related to the risk of type 2 diabetes mellitus (T2DM) through upregulated GRK5 mRNA expression. Rs10886471 is located in the intron region of GRK5. However, the mechanism
P Kunapuli et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(12), 5588-5592 (1993-06-15)
Guanine nucleotide binding protein (G-protein)-coupled receptor kinases (GRKs) specifically phosphorylate the agonist-occupied form of G-protein-coupled receptors such as the beta 2-adrenergic receptor and rhodopsin. The best characterized members of this family include the beta-adrenergic receptor kinase (beta ARK) and rhodopsin
Jie Fan et al.
Nature medicine, 9(3), 315-321 (2003-02-20)
Polymorphonuclear leukocytes (PMNs) are critical effector cells of the innate immune system that protect the host by migrating to inflammatory sites and killing pathogenic microbes. We addressed the role of chemokine receptor desensitization induced by G-protein-coupled receptor kinases (GRKs) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service