Skip to Content
Merck
All Photos(1)

Key Documents

G7509

Sigma-Aldrich

GABase from Pseudomonas fluorescens

lyophilized powder, Protein ≥30 % by biuret

Synonym(s):

GABase Enzyme, GABase from Pseudomonas, Pseudomonas GABase

Sign Into View Organizational & Contract Pricing

Select a Size

10 UNITS
$690.00
25 UNITS
$1,520.00

$690.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 UNITS
$690.00
25 UNITS
$1,520.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

$690.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

biological source

Pseudomonas fluorescens

Quality Level

Assay

≥30  % (biuret)

form

lyophilized powder

specific activity

≥0.5 units/mg protein

composition

Protein, ≥30% biuret

technique(s)

inhibition assay: suitable

storage temp.

−20°C

General description

Research area: Neuroscience


GABase is a commercially available mixture composed of γ-aminobutyric acid aminotransferase (GABGT) and succinic semialdehyde dehydrogenase (SSDH) obtained from Pseudomonas fluorescens.[1][2]

Application

GABase from Pseudomonas fluorescens has been used for GABase assay to quantify the extracellular and intracellular concentration of GABA (γ-aminobutyric acid).[1][2]

Biochem/physiol Actions

GABase catalyzes the conversion of γ-aminobutyric acid (GABA) to succinic acid in the central nervous system. It is used to measure GABA levels in various biological samples. p-Hydroxybenzaldehyde (HBA) is known to be a potential inhibitor on GABase.[3]

Unit Definition

One unit will convert 1.0 μmole of γ-aminobutyric acid (GABA) to succinic semialdehyde and then to succinate per min with a stoichiometric reduction of 1.0 μmole of NADP+ at pH 8.6 at 25 °C.

Physical form

Partially purified lyophilized powder containing buffer salts and stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingu Gordon Park et al.
Journal of enzyme inhibition and medicinal chemistry, 36(1), 2016-2024 (2021-09-14)
Many studies have focussed on modulating the activity of γ-aminobutyric acid transaminase (GABA-T), a GABA-catabolizing enzyme, for treating neurological diseases, such as epilepsy and drug addiction. Nevertheless, human GABA-T synthesis and purification have not been established. Thus, biochemical and drug
Kimon-Andreas G Karatzas et al.
Applied and environmental microbiology, 76(11), 3529-3537 (2010-04-20)
It is well established that the glutamate decarboxylase (GAD) system is central to the survival of Listeria monocytogenes at low pH, both in acidic foods and within the mammalian stomach. The accepted model proposes that under acidic conditions extracellular glutamate
Structural and Mechanistic Basis for the Inactivation of Human Ornithine Aminotransferase by (3 S,4 S)-3-Amino-4-fluorocyclopentenecarboxylic Acid
Shen S, et al.
Molecules (Basel), 28(3) (2023)
Nicolaie Moldovan et al.
Sensors and actuators. B, Chemical, 337 (2022-05-24)
Glutamate (GLU) and gamma-aminobutyric acid (GABA) are neurotransmitters (NTs) with an essential role in signal transmission in the brain. Brain disorders, such as epilepsy, Alzheimer's and Parkinson's diseases, and traumatic brain injury can be linked to imbalances in the GLU-GABA
A Vidal-Cros et al.
The Biochemical journal, 229(3), 675-678 (1985-08-01)
L-threo-3-Fluoroglutamate and L-erythro-3-fluoroglutamate were tested with glutamate decarboxylase from Escherichia coli. Both isomers were substrates: the threo isomer was decarboxylated into optically active 4-amino-3-fluorobutyrate, whereas the erythro isomer lost the fluorine atom during the reaction, yielding succinic semialdehyde after hydrolysis

Questions

1–4 of 4 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How do I solubilize Product No. G7509, GABase from Pseudomonas fluorescens?

    1 answer
    1. Sigma recommends to dissolve the product with 75 mM phosphate buffer, pH 7.2, containing 25% (v/v) glycerol to a concentration of 2-5 units per mL. Keep the solution in an ice bath while using and store solution frozen. This solution may be stored frozen and thawed several times with only slight loss of activity.

      Helpful?

  3. Are there any literature references that cite use of the Product No. G7509, GABase from Pseudomonas fluorescens?

    1 answer
    1. The product is cited in the reference Sulaimana, Saba A.J., et al., J of Enz Inhib and Med Chem. 18(4), 297-301 (2003).

      Helpful?

  4. What is the shelf life of the Product No.G7509, GABase from Pseudomonas fluorescens?

    1 answer
    1. Sigma recommends retesting the product 3 years after its release date. The retest date will appear on the lot specific Certificate of Analysis for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service