Skip to Content
Merck
All Photos(1)

Key Documents

G6125

Sigma-Aldrich

Glucose Oxidase from Aspergillus niger

Type II, ≥10,000 units/g solid (without added oxygen)

Synonym(s):

β-D-Glucose:oxygen 1-oxidoreductase, G.Od., GOx

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

type

Type II

Quality Level

form

powder

specific activity

≥10,000 units/g solid (without added oxygen)

mol wt

160 kDa

foreign activity

amylase ≤0.5%
catalase ≤2 Sigma units/mg solid
galactose oxidase ≤3%
glycogenase ≤0.5%
invertase ≤0.5%
maltase ≤2%

storage temp.

−20°C

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6-/m1/s1

InChI key

WQZGKKKJIJFFOK-VFUOTHLCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Weight: 160 kDa (gel filtration)
pI: 4.2
Extinction coefficient: E1% = 16.7 (280 nm)

Glucose oxidase from Aspergillus niger is a dimer consisting of 2 equal subunits with a molecular mass of 80 kDa each. Each subunit contains one flavin adenine dinulceotide moiety and one iron. The enzyme is a glycoprotein containing ~16% neutral sugar and 2% amino sugars. The enzyme also contains 3 cysteine residues and 8 potential sites for N-linked glycosylation.

Glucose oxidase is capable of oxidizing D-aldohexoses, monodeoxy-D-glucoses, and methyl-D-glucoses at varying rates.

The pH optimum for glucose oxidase is 5.5, while it has a broad activity range of pH 4-7. Glucose oxidase is specific for β-D-glucose with a KM of 33-110 mM.

Glucose oxidase does not require any activators, but it is inhibited by Ag+, Hg2+, Cu2+, phenylmercuric acetate, and p-chloromercuribenzoate. It is not inhibited by the nonmetallic SH reagents: N-ethylmaleimide, iodoacetate, and iodoacetamide.

Glucose oxidase can be utilized in the enzymatic determination of D-glucose in solution. As glucose oxidase oxidizes β-D-glucose to D-gluconolactate and hydrogen peroxide, horseradish peroxidase is often used as the coupling enzyme for glucose determination. Although glucose oxidase is specific for β-D-glucose, solutions of D-glucose can be quantified as α-D-glucose will mutorotate to β-D-glucose as the β-D-glucose is consumed by the enzymatic reaction.

Application

Glucose oxidase is widely used in the food and pharmaceutical industries as well as a major component of glucose biosensors.

Biochem/physiol Actions

Glucose oxidase catalyses the oxidation of β-d-glucose to d-glucono-β-lactone and hydrogen peroxide, with molecular oxygen as an electron acceptor.

Caution

Some loss of activity may occur after more than 3 days at room temperature. This product may be shipped with or without dry ice.

Linkage

This preparation is formulated from Type VII, G2133, by addition of potassium gluconate.

Unit Definition

One unit will oxidize 1.0 μmole of β-D-glucose to D-gluconolactone and H2O2 per min at pH 5.1 at 35 °C, equivalent to an O2 uptake of 22.4 μl per min. If the reaction mixture is saturated with oxygen, the activity may increase by up to 100%.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Na et al.
The Journal of comparative neurology, 527(15), 2488-2511 (2019-03-20)
The mammalian cerebellar cortex is compartmentalized, both anatomically and histochemically, into multiple parasagittal bands. To characterize the multiple zonal patterns of pontocerebellar mossy fiber projection, single neurons in the basilar pontine nucleus (BPN) were labeled by injecting biotinylated dextran amine
Charlotte F Kelley et al.
eLife, 9 (2020-07-14)
Focal adhesions (FA) are large macromolecular assemblies which help transmit mechanical forces and regulatory signals between the extracellular matrix and an interacting cell. Two key proteins talin and vinculin connecting integrin to actomyosin networks in the cell. Both proteins bind
Malgorzata Pierzchalska et al.
BioTechniques, 52(5), 307-315 (2012-05-15)
Investigating intestinal physiology in vitro remains challenging due to the lack of an effective primary enterocyte culture system. Recently developed protocols for growing organoids containing crypts and villus from adult mouse intestinal epithelium in Matrigel present an attractive alternative to
Riin Kont et al.
Nature communications, 11(1), 5786-5786 (2020-11-15)
Lytic polysaccharide monooxygenases (LPMOs) are widely distributed in Nature, where they catalyze the hydroxylation of glycosidic bonds in polysaccharides. Despite the importance of LPMOs in the global carbon cycle and in industrial biomass conversion, the catalytic properties of these monocopper
Timothy M Locke et al.
Frontiers in cellular neuroscience, 14, 228-228 (2020-08-28)
Tyrosine hydroxylase (Th) expression has previously been reported in Purkinje cells (PCs) of rodents and humans, but its role in the regulation of behavior is not understood. Catecholamines are well known for facilitating cognitive behaviors and are expressed in many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service