Skip to Content
Merck
All Photos(1)

Key Documents

G5048

Sigma-Aldrich

Geranylgeranylacetone

Synonym(s):

6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one, mixture of (5E,9E,13E) and (5Z,9E,13E) isomers, GGA, Selbex, Teprenone, UNII-S8S8451A4O

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$108.00
50 MG
$435.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$108.00
50 MG
$435.00

About This Item

Empirical Formula (Hill Notation):
C23H38O
CAS Number:
Molecular Weight:
330.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

oil

storage condition

protect from light

color

clear

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\CCC(C)=O

InChI

1S/C23H38O/c1-19(2)11-7-12-20(3)13-8-14-21(4)15-9-16-22(5)17-10-18-23(6)24/h11,13,15,17H,7-10,12,14,16,18H2,1-6H3/b20-13+,21-15+,22-17+

InChI key

HUCXKZBETONXFO-NJFMWZAGSA-N

Application

Geranylgeranylacetone has been used as an inducer of heat shock protein 70 (HSP70) to analyze its protective effects against cerebral ischemia/reperfusion (I/R).[1]

Biochem/physiol Actions

Geranylgeranylacetone can induce expression of HSP70, HSPB8, and HSPB1. Induction of HSP70 expression is protective against the development of various diseases, such as inflammatory bowel disease, hypoxic/ischemic brain injury and spinal and bulbar muscular atrophy (cytoprotective and anti-inflammatory effects). Reports indicate that GGA protects against NSAID-induced gastric and intestinal lesions by induction of HSP70 expression. Other studies have shown that GGA induces expression of HSPB8 and HSPB1 and reduces the formation of amyloid oligomers as well as insoluble aggregates in HSPB5 R120G TG mice.
Geranylgeranylacetone is an acyclic polyisoprenoid that has been studied to exhibit protective effects against reperfusion injury, inflammation and organ transplantation. It is an anti-ulcer agent that is involved in the protection of gastric mucosa.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoko Ishii et al.
Investigative ophthalmology & visual science, 44(5), 1982-1992 (2003-04-26)
To study the effects of geranylgeranylacetone (GGA) on the expression of inducible (HSP72) and constitutive (HSC70) heat shock proteins (HSPs) on retinal ganglion cells (RGCs) in a rat model of glaucoma. Adult Wistar rats were given intraperitoneal injections of GGA
Kylie Kavanagh et al.
American journal of physiology. Endocrinology and metabolism, 300(5), E894-E901 (2011-02-18)
We evaluated heat shock protein 70 (HSP70) changes in diabetes mellitus (DM) in a nonhuman primate model. To this end, two studies were conducted in DM vervet monkeys. 1) Normal control and streptozotocin-induced DM monkeys (Stz-DM) that were differentiated into
Tetsuro Marunouchi et al.
European journal of pharmacology, 730, 140-147 (2014-03-19)
The mechanisms underlying mitochondrial impairment in the failing heart are not yet clear. In a previous study, we found that the levels of small heat shock proteins (HSP) such as mitochondrial HSPB1 and HSPB8 in the failing heart following myocardial
Noritaka Fujimura et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(1), 153-160 (2011-10-15)
Geranylgeranylacetone (GGA) induces expression of heat shock protein 90 (Hsp90), an adaptor molecule for assembly of endothelial nitric oxide synthase (eNOS) phosphorylation complex. The purpose of this study was to determine whether GGA enhances Hsp90 expression and augments endothelium-dependent vasodilation
Fu-Cheng Luo et al.
Free radical biology & medicine, 52(7), 1218-1227 (2012-01-31)
There are few efficacious interventions to combat morphine dependence. Thioredoxin-1 (Trx-1) and heat shock protein 70 (Hsp70) are emerging as important modulators of neuronal function. They have been shown to be involved in cellular protective mechanisms against a variety of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service