Skip to Content
Merck
All Photos(1)

Documents

G5044

Sigma-Aldrich

GR 32191B

≥98% (HPLC), solid

Synonym(s):

(4Z)-7-[(1R,2R,3S,5S)-5-([1,1′-Biphenyl]-4-ylmethoxy)-3-hydroxy-2-(1-piperidinyl)cyclopentyl]-4-heptenoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40NO4Cl
CAS Number:
Molecular Weight:
514.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

off-white

mp

127-128 °C

solubility

H2O: ~16 mg/mL

originator

GlaxoSmithKline

shipped in

wet ice

storage temp.

−20°C

SMILES string

Cl.O[C@H]1C[C@H](OCc2ccc(cc2)-c3ccccc3)[C@H](CC\C=C/CCC(O)=O)[C@H]1N4CCCCC4

InChI

1S/C30H39NO4.ClH/c32-27-21-28(35-22-23-15-17-25(18-16-23)24-11-5-3-6-12-24)26(13-7-1-2-8-14-29(33)34)30(27)31-19-9-4-10-20-31;/h1-3,5-6,11-12,15-18,26-28,30,32H,4,7-10,13-14,19-22H2,(H,33,34);1H/b2-1-;/t26-,27-,28-,30+;/m0./s1

InChI key

ZYOBZRTZRQKKNC-UGNABIHOSA-N

Biochem/physiol Actions

Selective TP prostanoid receptor antagonist. Inhibits U-46619-induced contraction of guinea pig trachea with IC50=1.8 nM.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Manufactured and sold under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service