Skip to Content
Merck
All Photos(1)

Key Documents

G4013

Sigma-Aldrich

Glutathione Peroxidase from human erythrocytes

lyophilized powder, ≥30 units/mg protein

Synonym(s):

GSH-Px, Glutathione:hydrogen-peroxide oxido-reductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥30 units/mg protein

composition

Protein, 13-25% modified Warburg-Christian

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPX1(2876)

General description

GPX1 (glutathione peroxidase 1) belongs to the four membered family of GPXs, including GPX2, GPX3 and GPX4. It is a selenocysteine-containing protein. GPX1 is the major isoform of GPX enzyme and is ubiquitously present intracellularly.
Note: At the reported pH optimum of 8.8, we have found the activity to be approx. 10 times that at pH 7.0. However, to remain consistent with literature and avoid complications arising from non-enzymatic oxidation of GSH, our unit is defined at pH 7.0.

Application

Glutathione peroxidase (GPX) from human erythrocytes has been used for the determination of the ability of peroxides to inactivate GPX in epithelial sheets obtained from cutaneous squamous cell carcinoma (CSCC) or solar keratoses (SK).

Biochem/physiol Actions

GPX1 (glutathione peroxidase 1) catalyzes the conversion of multiple organic and inorganic peroxides, by utilizing reduced glutathione (GSH) as an electron donor. This enzyme is also responsible for the removal of organic and inorganic peroxides.

Unit Definition

One unit will catalyze the oxidation by H2O2 of 1.0 μmole of reduced glutathione to oxidized glutathione per min at pH 7.0 at 25 °C.

Physical form

Lyophilized powder containing 25% sucrose, 3% dithiothreitol, and sodium phosphate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Walshe et al.
Cancer research, 67(10), 4751-4758 (2007-05-19)
Cutaneous squamous cell carcinomas (CSCC) are a common malignancy of keratinocytes that arise in sites of the skin exposed to excessive UV radiation. In the present study, we show that human SCC cell lines, preneoplastic solar keratoses (SK), and CSCC
Tengku Ahbrizal Farizal Tengku Ahmad et al.
Molecules (Basel, Switzerland), 18(2), 2200-2211 (2013-02-26)
The present study was designed to determine the radioprotective effects of Malaysian Gelam honey on gene expression and enzyme activity of superoxide dismutase (SOD), catalase (CAT) and glutathione peroxidase (GPx) of human diploid fibroblasts (HDFs) subjected to gamma-irradiation. Six groups
O L Gostukhina et al.
Zhurnal evoliutsionnoi biokhimii i fiziologii, 48(6), 542-547 (2013-02-14)
The work studied the content of carotinoids, the state of antioxidant (AO) enzymatic complex, and intensity of lipid peroxidation in tissues (hepatopancreas, gill, foot) of the Black Sea bivalve mollusc Anadara inaequivalvis. Tissues with a high content of the pigment
Hai-Dong Yao et al.
The Journal of nutrition, 143(5), 613-619 (2013-03-22)
Dietary selenium (Se) deficiency causes muscular dystrophy in various species, but the molecular mechanism remains unclear. Our objectives were to investigate: 1) if dietary Se deficiency induced different amounts of oxidative stress, lipid peroxidation, and cell apoptosis in 3 skeletal
Bing-Lin Zhang et al.
Environmental toxicology and chemistry, 32(6), 1420-1425 (2013-02-19)
The effect of sodium chloride (NaCl) on cadmium (Cd) uptake, translocation, and oxidative stress was investigated using 2 tobacco cultivars differing in Cd tolerance. The growth inhibition of the tobacco plants exposed to Cd toxicity was in part alleviated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service